Gene Transcripts

Gene Symbol
Klrg1
Entrez Gene ID
58975
Gene Name
killer cell lectin-like receptor subfamily G, member 1
Gene Aliases
MAFA
Chromosome Location
Chr. 4: 155038936 - 155051444 on Build Rnor_6.0
UniGene ID
Rn.10256
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031649.2 NP_113837.1 1 14
XM_006237313.2 XP_006237375.1 1 82
GenBank mRNA X79812.1 - 1 67

View Full Product Details