Gene Transcripts

Gene Symbol
Renbp
Entrez Gene ID
81759
Gene Name
renin binding protein
Gene Aliases
Chromosome Location
Chr. X: 156854488 - 156863544 on Build Rnor_6.0
UniGene ID
Rn.11116
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031095.1 NP_112357.1 7, 8 934
XM_006229615.2 XP_006229677.1 8, 9 1056
GenBank mRNA D10233.1 - 7, 8 934

View Full Product Details