Gene Transcripts

Gene Symbol
Plcl1
Entrez Gene ID
84587
Gene Name
phospholipase C-like 1
Gene Aliases
Chromosome Location
Chr. 9: 62016112 - 62349244 on Build Rnor_6.0
UniGene ID
Rn.10684
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053456.1 NP_445908.1 2 1411
XM_008767141.1 XP_008765363.1 1 5192
GenBank mRNA D45920.1 - 2 1411

View Full Product Details