Gene Transcripts

Gene Symbol
Ppp1r2
Entrez Gene ID
192361
Gene Name
protein phosphatase 1, regulatory (inhibitor) subunit 2
Gene Aliases
IPP-2
Chromosome Location
Chr. 11: 72748789 - 72759946 on Build Rnor_6.0
UniGene ID
Rn.4016
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138823.2 NP_620178.1 170
GenBank mRNA BC092194.1 - 170
S79213.1 - 185

View Full Product Details