Gene Transcripts

Gene Symbol
Tnfrsf8
Entrez Gene ID
25069
Gene Name
tumor necrosis factor receptor superfamily, member 8
Gene Aliases
Cd30
Chromosome Location
Chr. 5: 163186349 - 163232139 on Build Rnor_6.0
UniGene ID
Rn.11322
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019135.1 NP_062008.1 3 284
GenBank mRNA D42117.1 - 3 284

View Full Product Details