Gene Transcripts

Gene Symbol
Hoxa1
Entrez Gene ID
25607
Gene Name
homeo box A1
Gene Aliases
Chromosome Location
Chr. 4|NW_007906169: 25763 - 28587 on Build Rnor_6.0
UniGene ID
Rn.228589
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013075.1 NP_037207.1 1 626
XM_008773708.1 XP_008771930.1 2 539
GenBank mRNA U93092.1 - 1 626

Gene Symbol
NEWGENE_2812
Entrez Gene ID
103690132
Gene Name
homeobox A1
Gene Aliases
Hoxa1
Chromosome Location
Chr. 4: 82124358 - 82127182 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008762937.1 XP_008761159.1 1 742
XM_008762938.1 XP_008761160.1 2 539

View Full Product Details