Gene Transcripts

Gene Symbol
Tspy26
Entrez Gene ID
296280
Gene Name
testis specific protein, Y-linked 26
Gene Aliases
RGD1310192, Tspyl3
Chromosome Location
Chr. 3: 148704896 - 148706972 on Build Rnor_6.0
UniGene ID
Rn.125787
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106527.2 NP_001099997.1 1 789
GenBank mRNA BC168711.1 - 1 789

View Full Product Details