Gene Transcripts

Gene Symbol
Fbxo41
Entrez Gene ID
312504
Gene Name
F-box protein 41
Gene Aliases
RGD1566130
Chromosome Location
Chr. 4: 117255143 - 117285228 on Build Rnor_6.0
UniGene ID
Rn.147439
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107871.1 NP_001101341.1 7 2267

View Full Product Details