Gene Transcripts

Gene Symbol
Hmg20a
Entrez Gene ID
315689
Gene Name
high mobility group 20A
Gene Aliases
RGD1564760
Chromosome Location
Chr. 8: 61080103 - 61157613 on Build Rnor_6.0
UniGene ID
Rn.9628
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108150.1 NP_001101620.1 4 602
XM_006243117.2 XP_006243179.1 4 4159

View Full Product Details