Gene Transcripts

Gene Symbol
Olr309
Entrez Gene ID
293607
Gene Name
olfactory receptor 309
Gene Aliases
ratchr1-200857727-200858659_ORF
Chromosome Location
Chr. 1: 213334956 - 213335888 on Build Rnor_6.0
UniGene ID
Rn.141729
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000242.1 NP_001000242.1 1 470

View Full Product Details