Gene Transcripts

Gene Symbol
RGD1564804
Entrez Gene ID
313551
Gene Name
similar to chromosome 1 open reading frame 50
Gene Aliases
Chromosome Location
Chr. 5: 138274907 - 138279285 on Build Rnor_6.0
UniGene ID
Rn.7322
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107969.1 NP_001101439.1 4 407
GenBank mRNA FQ209773.1 - 4 433

View Full Product Details