Gene Transcripts

Gene Symbol
Snap23
Entrez Gene ID
64630
Gene Name
synaptosomal-associated protein 23
Gene Aliases
Chromosome Location
Chr. 3: 112333972 - 112362960 on Build Rnor_6.0
UniGene ID
Rn.14789
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022689.2 NP_073180.1 7 756
GenBank mRNA AF052596.1 - 608
BC127494.1 - 7 756 8361815

View Full Product Details