Gene Transcripts

Gene Symbol
Dusp5
Entrez Gene ID
171109
Gene Name
dual specificity phosphatase 5
Gene Aliases
Cpg21
Chromosome Location
Chr. 1: 274245184 - 274258595 on Build Rnor_6.0
UniGene ID
Rn.10877
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133578.1 NP_598262.1 2 682
GenBank mRNA AF013144.1 - 2 682

View Full Product Details