Gene Transcripts

Gene Symbol
Chrnb3
Entrez Gene ID
171131
Gene Name
cholinergic receptor, nicotinic, beta 3 (neuronal)
Gene Aliases
Chromosome Location
Chr. 16: 68876442 - 68913628 on Build Rnor_6.0
UniGene ID
Rn.9694
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133597.1 NP_598281.1 4 485
GenBank mRNA AY574259.1 - 4 308
J04636.1 - 4 485

View Full Product Details