Gene Transcripts

Gene Symbol
Spon2
Entrez Gene ID
171569
Gene Name
spondin 2, extracellular matrix protein
Gene Aliases
Chromosome Location
Chr. 14: 82818735 - 82824529 on Build Rnor_6.0
UniGene ID
Rn.6699
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138533.3 NP_612542.1 4 684
GenBank mRNA BC078734.1 - 4 684

Gene Symbol
LOC100910790
Entrez Gene ID
100910790
Gene Name
spondin-2-like
Gene Aliases
Chromosome Location
Chr. 14: 82867099 - 82889642 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008770410.1 XP_008768632.1 800

View Full Product Details