Gene Transcripts

Gene Symbol
Ptgis
Entrez Gene ID
25527
Gene Name
prostaglandin I2 (prostacyclin) synthase
Gene Aliases
PGIS
Chromosome Location
Chr. 3: 163950746 - 163986129 on Build Rnor_6.0
UniGene ID
Rn.73051
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031557.2 NP_113745.1 3 488
GenBank mRNA BC061814.1 - 3 488
U53855.1 - 3 419

View Full Product Details