Gene Transcripts

Gene Symbol
Cpe
Entrez Gene ID
25669
Gene Name
carboxypeptidase E
Gene Aliases
CARBE, Cph
Chromosome Location
Chr. 16: 26906716 - 27014811 on Build Rnor_6.0
UniGene ID
Rn.7149
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013128.1 NP_037260.1 1 361
GenBank mRNA BC085762.1 - 1 360
J04625.1 - 1 316
M31602.1 - 1 332
X51406.1 - 1 361

View Full Product Details