Gene Transcripts

Gene Symbol
Pnrc1
Entrez Gene ID
286988
Gene Name
proline-rich nuclear receptor coactivator 1
Gene Aliases
Prol2
Chromosome Location
Chr. 5: 48501472 - 48504511 on Build Rnor_6.0
UniGene ID
Rn.10967
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173322.1 NP_775444.1 1 406
GenBank mRNA FQ214116.1 - 1 448
U61729.1 - 1 406

View Full Product Details