Gene Transcripts

Gene Symbol
Tnnc1
Entrez Gene ID
290561
Gene Name
troponin C type 1 (slow)
Gene Aliases
Tncc
Chromosome Location
Chr. 16: 7292207 - 7295425 on Build Rnor_6.0
UniGene ID
Rn.15460
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034105.1 NP_001029277.1 4 293
XM_008770987.1 XP_008769209.1 5 437
GenBank mRNA DQ062205.1 - 4 293

View Full Product Details