Gene Transcripts

Gene Symbol
Tubb4a
Entrez Gene ID
29213
Gene Name
tubulin, beta 4A class IVa
Gene Aliases
Tubb4
Chromosome Location
Chr. 9: 9961020 - 9968420 on Build Rnor_6.0
UniGene ID
Rn.195342
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080882.1 NP_543158.1 2, 3 276
GenBank mRNA BC168216.1 - 2, 3 281
CO395990.1 - 2, 3 279

View Full Product Details