Gene Transcripts

Gene Symbol
RGD1307554
Entrez Gene ID
292739
Gene Name
similar to CG16812-PA
Gene Aliases
Chromosome Location
Chr. 1: 84361720 - 84388765 on Build Rnor_6.0
UniGene ID
Rn.106026
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109664.1 NP_001103134.1 10 1275
XM_008759118.1 XP_008757340.1 9 1561

View Full Product Details