Gene Transcripts

Gene Symbol
Aoc3
Entrez Gene ID
29473
Gene Name
amine oxidase, copper containing 3
Gene Aliases
SSAO
Chromosome Location
Chr. 10: 89251370 - 89259313 on Build Rnor_6.0
UniGene ID
Rn.198327
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031582.2 NP_113770.2 1 1219
GenBank mRNA AB195675.1 - 1 1224
BC100613.1 - 1 1219 5598211

View Full Product Details