Gene Transcripts

Gene Symbol
Trim29
Entrez Gene ID
300656
Gene Name
tripartite motif-containing 29
Gene Aliases
Chromosome Location
Chr. 8: 47674321 - 47699065 on Build Rnor_6.0
UniGene ID
Rn.9452
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106815.1 NP_001100285.1 5 1470

View Full Product Details