Gene Transcripts

Gene Symbol
Spg21
Entrez Gene ID
300791
Gene Name
spastic paraplegia 21 homolog (human)
Gene Aliases
Chromosome Location
Chr. 8: 70994532 - 71022302 on Build Rnor_6.0
UniGene ID
Rn.19459
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001006987.1 NP_001006988.1 5, 6 677
XM_006243251.2 XP_006243313.1 5, 6 708
GenBank mRNA BC083760.1 - 5, 6 677

View Full Product Details