Gene Transcripts

Gene Symbol
Col9a1
Entrez Gene ID
305104
Gene Name
collagen, type IX, alpha 1
Gene Aliases
Chromosome Location
Chr. 9: 30419001 - 30502307 on Build Rnor_6.0
UniGene ID
Rn.90726
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100842.1 NP_001094312.1 27 2040
GenBank mRNA DY316977.1 - 1 17

View Full Product Details