Gene Transcripts

Gene Symbol
Ell2
Entrez Gene ID
309918
Gene Name
elongation factor RNA polymerase II 2
Gene Aliases
Chromosome Location
Chr. 2: 2456189 - 2525590 on Build Rnor_6.0
UniGene ID
Rn.105837
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008760603.1 XP_008758825.1 3 961
XM_008760604.1 XP_008758826.1 3 247

View Full Product Details