Gene Transcripts

Gene Symbol
Olr1132
Entrez Gene ID
405344
Gene Name
olfactory receptor 1132
Gene Aliases
Chromosome Location
Chr. 8: 19019410 - 19020378 on Build Rnor_6.0
UniGene ID
Rn.143605
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000986.1 NP_001000986.1 1 266

View Full Product Details