Gene Transcripts

Gene Symbol
Vom1r107
Entrez Gene ID
494260
Gene Name
vomeronasal 1 receptor 107
Gene Aliases
V1rg4
Chromosome Location
Chr. 7: 18068060 - 18069043 on Build Rnor_6.0
UniGene ID
Rn.129737
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008908.1 NP_001008908.1 1 471
GenBank mRNA AY510300.1 - 1 471

View Full Product Details