Gene Transcripts

Gene Symbol
Tspan33
Entrez Gene ID
500065
Gene Name
tetraspanin 33
Gene Aliases
RGD1560915
Chromosome Location
Chr. 4: 56981283 - 57005866 on Build Rnor_6.0
UniGene ID
Rn.1696
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109227.1 NP_001102697.1 7 840
XM_006236227.2 XP_006236289.1 7 826

View Full Product Details