Gene Transcripts

Gene Symbol
Cd300ld
Entrez Gene ID
100361633
Gene Name
Cd300 molecule-like family member D
Gene Aliases
Chromosome Location
Chr. 10: 104847315 - 104895630 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_002727841.3 XP_002727887.2 2 419
XM_008768422.1 XP_008766644.1 2 652
XM_008768423.1 XP_008766645.1 2 645
XM_008768424.1 XP_008766646.1 2 652

View Full Product Details