Gene Transcripts

Gene Symbol
Arpp19
Entrez Gene ID
60336
Gene Name
cAMP-regulated phosphoprotein 19
Gene Aliases
Arpp-19
Chromosome Location
Chr. 8: 81949582 - 81972054 on Build Rnor_6.0
UniGene ID
Rn.112577
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031660.1 NP_113848.1 3 313
GenBank mRNA AJ005982.1 - 3 313

View Full Product Details