Gene Transcripts

Gene Symbol
Fhit
Entrez Gene ID
60398
Gene Name
fragile histidine triad
Gene Aliases
Chromosome Location
Chr. 15: 15697292 - 16862873 on Build Rnor_6.0
UniGene ID
Rn.45598
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021774.1 NP_068542.1 6 576
GenBank mRNA AF170064.1 - 6 576

View Full Product Details