Gene Transcripts

Gene Symbol
Nmu
Entrez Gene ID
63887
Gene Name
neuromedin U
Gene Aliases
Chromosome Location
Chr. 14: 34353683 - 34381968 on Build Rnor_6.0
UniGene ID
Rn.47720
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022239.2 NP_071575.1 3, 4 342
XM_006250872.2 XP_006250934.1 3, 4 1162
GenBank mRNA AJ510132.1 - 2, 3 101
M94555.1 - 3, 4 342

View Full Product Details