Gene Transcripts

Gene Symbol
Scamp4
Entrez Gene ID
65170
Gene Name
secretory carrier membrane protein 4
Gene Aliases
Chromosome Location
Chr. 7: 11969720 - 11982141 on Build Rnor_6.0
UniGene ID
Rn.7309
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031725.1 NP_113913.1 7 751
GenBank mRNA AF285631.1 - 7 751

View Full Product Details