Gene Transcripts

Gene Symbol
Pcca
Entrez Gene ID
687008
Gene Name
propionyl CoA carboxylase, alpha polypeptide
Gene Aliases
Chromosome Location
Chr. 15: 108960509 - 109306879 on Build Rnor_6.0
UniGene ID
Rn.6033
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019330.1 NP_062203.1 17, 18 1658
XM_008770973.1 XP_008769195.1 17, 18 1598
GenBank mRNA CK601565.1 - 563
CO574538.1 - 3, 4 297

View Full Product Details