Gene Transcripts

Gene Symbol
Taldo1
Entrez Gene ID
83688
Gene Name
transaldolase 1
Gene Aliases
Chromosome Location
Chr. 1: 214375555 - 214385886 on Build Rnor_6.0
UniGene ID
Rn.3136
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031811.3 NP_113999.2 4 430
GenBank mRNA AF069306.1 - 4 416
BC059126.1 - 4 434 6889440
FQ213939.1 - 4 442

View Full Product Details