Gene Transcripts

Gene Symbol
Tnfaip6
Entrez Gene ID
84397
Gene Name
tumor necrosis factor alpha induced protein 6
Gene Aliases
Tnfip6
Chromosome Location
Chr. 3: 37545238 - 37564704 on Build Rnor_6.0
UniGene ID
Rn.195358
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053382.1 NP_445834.1 1, 2 119
GenBank mRNA BC158666.1 - 1, 2 119
FQ228771.1 - 1, 2 181
FQ228783.1 - 1, 2 181

View Full Product Details