Gene Transcripts

Gene Symbol
Cdhr1
Entrez Gene ID
93662
Gene Name
cadherin-related family member 1
Gene Aliases
Pcdh21
Chromosome Location
Chr. 16: 14328161 - 14348049 on Build Rnor_6.0
UniGene ID
Rn.19780
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053572.1 NP_446024.1 10 1151
GenBank mRNA AB053449.1 - 10 1151

View Full Product Details