Gene Transcripts

Gene Symbol
FGF10
Entrez Gene ID
2255
Gene Name
fibroblast growth factor 10
Gene Aliases
-
Chromosome Location
Chr.5: 44301655 - 44389780 on Build GRCh38
Uni Gene
Hs.248049
Interrogated Sequence Translated Protein
RefSeq NM_004465.1 NP_004456.1
XM_005248264.3 XP_005248321.1
GenBank mRNA AB002097.1 -
AF411527.1 -
AF508782.1 -
AK023508.1 -
AK315314.1 -
BC069561.1 -
BC105021.1 -
BC105023.1 -
BC143772.1 -
CR541665.1 -
CR541688.1 -
ENST00000264664.0 -
GQ351295.1 -
U67918.1 -

View Full Product Details