Gene Transcripts

Gene Symbol
IFNB1
Entrez Gene ID
3456
Gene Name
interferon beta 1
Gene Aliases
IFB, IFF, IFN-beta, IFNB
Chromosome Location
Chr.9: 21077105 - 21077963 on Build GRCh38
Uni Gene
Hs.93177
Interrogated Sequence Translated Protein
RefSeq NM_002176.3 NP_002167.1
GenBank mRNA AB451323.1 -
AB451452.1 -
AB451491.1 -
BC069314.1 -
BC096150.3 -
BC096151.2 -
BC096152.2 -
BC096153.2 -
ENST00000380232.0 -
M17373.2 -
M25460.1 -
M28622.1 -
V00546.1 -
V00547.1 -

View Full Product Details