Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_001079846.1 | NP_001073315.1 |
NM_004380.2 | NP_004371.2 | |
XM_005255124.4 | XP_005255181.1 | |
XM_005255125.4 | XP_005255182.1 | |
XM_006720848.3 | XP_006720911.1 | |
XM_011522381.2 | XP_011520683.1 | |
XM_011522382.2 | XP_011520684.1 | |
XM_017022944.1 | XP_016878433.1 | |
GenBank mRNA | AB210043.1 | - |
AK124320.1 | - | |
AK304771.1 | - | |
BC036486.1 | - | |
ENST00000262367.0 | - | |
U47741.1 | - | |
U85962.3 | - | |
U89354.1 | - | |
U89355.1 | - |