Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_000660.5 | NP_000651.3 |
XM_011527242.1 | XP_011525544.1 | |
GenBank mRNA | AI304490.1 | - |
AK291907.1 | - | |
AK307742.1 | - | |
AY820829.1 | - | |
BC000125.1 | - | |
BC001180.1 | - | |
BC022242.1 | - | |
BG115704.1 | - | |
BP267938.1 | - | |
BT007245.1 | - | |
DA935180.1 | - | |
ENST00000221930.0 | - | |
M38449.1 | - | |
X02812.1 | - |