Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_000358.2 | NP_000349.1 |
GenBank mRNA | AB209598.1 | - |
AK093916.1 | - | |
AK094055.1 | - | |
AK094581.1 | - | |
AK222833.1 | - | |
AK298603.1 | - | |
AK303643.1 | - | |
BC000097.2 | - | |
BC004972.1 | - | |
BC026352.1 | - | |
BT009820.1 | - | |
DA051240.1 | - | |
ENST00000442011.0 | - | |
KU178357.1 | - | |
KU178358.1 | - | |
M77349.1 | - |