Gene Transcripts

Gene Symbol
IKBKG
Entrez Gene ID
8517
Gene Name
inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase gamma
Gene Aliases
AMCBX1, FIP-3, FIP3, Fip3p, IKK-gamma, IKKAP1, IKKG, IMD33, IP, IP1, IP2, IPD2, NEMO, ZC2HC9
Chromosome Location
Chr.X: 154542240 - 154565046 on Build GRCh38
Uni Gene
Hs.43505
Interrogated Sequence Translated Protein
RefSeq NM_001099856.4 NP_001093326.2
NM_001099857.2 NP_001093327.1
NM_001145255.2 NP_001138727.1
NM_001321396.1 NP_001308325.1
NM_001321397.1 NP_001308326.1
NM_003639.4 NP_003630.1
GenBank mRNA AF062089.1 -
AF074382.1 -
AF091453.1 -
AF261086.1 -
AK000593.1 -
AK308366.1 -
AY114157.1 -
AY429546.1 -
AY429547.1 -
AY429548.1 -
BC000299.2 -
BC012114.1 -
BC046922.1 -
BC050612.2 -
BM473416.1 -
BT019621.1 -
CA916723.1 -
DC410130.1 -

View Full Product Details