Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_002646.3 | NP_002637.3 |
XM_005245257.2 | XP_005245314.1 | |
XM_005245258.3 | XP_005245315.1 | |
XM_011509630.2 | XP_011507932.1 | |
XM_011509631.2 | XP_011507933.1 | |
XM_011509633.2 | XP_011507935.1 | |
XM_017001473.1 | XP_016856962.1 | |
XM_017001474.1 | XP_016856963.1 | |
XM_017001475.1 | XP_016856964.1 | |
GenBank mRNA | AB210006.1 | - |
AK308153.1 | - | |
BC094741.1 | - | |
BC132876.1 | - | |
BC136645.1 | - | |
BC144342.1 | - | |
CR749201.1 | - | |
ENST00000367187.0 | - | |
F17716.1 | - | |
Y11312.1 | - | |
Y13892.1 | - |