Gene Transcripts

Gene Symbol
CCL13
Entrez Gene ID
6357
Gene Name
C-C motif chemokine ligand 13
Gene Aliases
CKb10, MCP-4, NCC-1, NCC1, SCYA13, SCYL1
Chromosome Location
Chr.17: 34356452 - 34358610 on Build GRCh38
Uni Gene
Hs.414629
Interrogated Sequence Translated Protein
RefSeq NM_005408.2 NP_005399.1
GenBank mRNA AJ001634.1 -
BC008621.1 -
BT007385.1 -
CR450337.1 -
ENST00000225844.0 -
U46767.1 -
U59808.1 -
X98306.1 -
Z77650.1 -
Z77651.1 -

View Full Product Details