Gene Transcripts

Gene Symbol
CTNNB1
Entrez Gene ID
1499
Gene Name
catenin beta 1
Gene Aliases
CTNNB, MRD19, armadillo
Chromosome Location
Chr.3: 41199451 - 41240448 on Build GRCh38
Uni Gene
Hs.476018
Interrogated Sequence Translated Protein
RefSeq NM_001098209.1 NP_001091679.1
NM_001098210.1 NP_001091680.1
NM_001904.3 NP_001895.1
XM_005264886.2 XP_005264943.1
XM_006712983.1 XP_006713046.1
XM_006712984.1 XP_006713047.1
XM_006712985.1 XP_006713048.1
XM_017005738.1 XP_016861227.1
GenBank mRNA AB062292.1 -
AB451264.1 -
AB451392.1 -
AK095242.1 -
AK289932.1 -
AK294778.1 -
AK296790.1 -
AK299954.1 -
AK314776.1 -
BC058926.1 -
BX648430.1 -
DA216720.1 -
ENST00000349496.0 -
U38440.1 -
X87838.1 -
Z19054.1 -

View Full Product Details