Gene Transcripts

Gene Symbol
TNFRSF11B
Entrez Gene ID
4982
Gene Name
TNF receptor superfamily member 11b
Gene Aliases
OCIF, OPG, PDB5, TR1
Chromosome Location
Chr.8: 118923557 - 118952144 on Build GRCh38
Uni Gene
Hs.81791
Interrogated Sequence Translated Protein
RefSeq NM_002546.3 NP_002537.3
GenBank mRNA AB002146.1 -
AF134187.1 -
AK223155.1 -
AK308524.1 -
AK313710.1 -
BC011023.1 -
BC030155.2 -
BM748946.1 -
BQ000897.1 -
CK903484.1 -
DA477521.1 -
ENST00000297350.0 -
U94332.1 -

View Full Product Details