Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_001007792.1 | NP_001007793.1 |
NM_001012331.1 | NP_001012331.1 | |
NM_002529.3 | NP_002520.2 | |
GenBank mRNA | AK126428.1 | - |
AK290759.1 | - | |
AK312704.1 | - | |
BC062580.1 | - | |
BC136554.1 | - | |
BC144239.1 | - | |
BM685020.1 | - | |
CR936794.1 | - | |
ENST00000524377.0 | - | |
KJ534902.1 | - | |
KJ534903.1 | - | |
M23102.1 | - |