Gene Transcripts

Gene Symbol
CREBBP
Entrez Gene ID
1387
Gene Name
CREB binding protein
Gene Aliases
CBP, KAT3A, RSTS
Chromosome Location
Chr.16: 3725054 - 3880727 on Build GRCh38
Uni Gene
Hs.459759
Interrogated Sequence Translated Protein
RefSeq NM_001079846.1 NP_001073315.1
NM_004380.2 NP_004371.2
XM_005255124.4 XP_005255181.1
XM_005255125.4 XP_005255182.1
XM_006720848.3 XP_006720911.1
XM_011522381.2 XP_011520683.1
XM_011522382.2 XP_011520684.1
XM_017022944.1 XP_016878433.1
GenBank mRNA AB210043.1 -
AK124320.1 -
AK304771.1 -
BC036486.1 -
ENST00000262367.0 -
U47741.1 -
U85962.3 -
U89354.1 -
U89355.1 -

View Full Product Details